siRNA Details
Gene Symbol: Tfrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022712.1 | NP_073203.1 | 9 | 1068 | ||
XM_006248462.2 | XP_006248524.1 | 9 | 1178 | |||
XM_006248463.2 | XP_006248525.1 | 9 | 1090 | |||
GenBank mRNA | FM033330.1 | - | 4 | 348 | ||
M58040.1 | - | 6 | 519 |
