siRNA Details
Gene Symbol: Tfrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022712.1 | NP_073203.1 | 3 | 224 | ||
XM_006248462.2 | XP_006248524.1 | 3 | 334 | |||
XM_006248463.2 | XP_006248525.1 | 3 | 246 | |||
GenBank mRNA | CB762571.1 | - | 3 | 236 | ||
CB809947.1 | - | 3 | 218 |

More Information
Other Species Targeted

Gene Ontology Categories:
