siRNA Details
Gene Symbol: Unc13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042579.1 | NP_001036044.1 | 26 | 2924 | ||
NM_022862.1 | NP_074053.1 | 17 | 4398 | |||
GenBank mRNA | AF159706.1 | - | 26 | 2924 | ||
U24071.1 | - | 17 | 4398 |
