siRNA Details
Gene Symbol: Pcdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022868.1 | NP_074059.1 | 3 | 2760 | ||
XM_006252375.2 | XP_006252437.1 | 3 | 3717 | |||
XM_008770897.1 | XP_008769119.1 | 4 | 3480 | |||
GenBank mRNA | AB026154.1 | - | 3 | 2760 |
