siRNA Details
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022955.1 | NP_075244.1 | 4020 | |||
GenBank mRNA | AB011532.1 | - | 4020 |
Gene Symbol: LOC100911486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764417.1 | XP_008762639.1 | 29 | 3681 |
