siRNA Details
Gene Symbol: Gabre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023091.1 | NP_075579.1 | 4 | 1994 | ||
GenBank mRNA | AF189262.1 | - | 4 | 1994 | ||
AF255385.1 | - | 1556 | ||||
AF255386.1 | - | 2 | 249 | |||
AF255387.1 | - | 2 | 249 | |||
AF255612.1 | - | 559 |
