siRNA Details
Gene Symbol: Myo1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023092.1 | NP_075580.2 | 29 | 3083 | ||
GenBank mRNA | X74800.1 | - | 29 | 3083 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myo1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023092.1 | NP_075580.2 | 29 | 3083 | ||
GenBank mRNA | X74800.1 | - | 29 | 3083 |