siRNA Details
Gene Symbol: Gapdhs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023964.1 | NP_076454.1 | 9, 10 | 1138 | ||
XM_006228790.2 | XP_006228852.1 | 9, 10 | 1508 | |||
XM_006228791.2 | XP_006228853.1 | 9, 10 | 1369 | |||
GenBank mRNA | AJ297631.1 | - | 9, 10 | 1138 | ||
BC161847.1 | - | 9, 10 | 1160 | 7112898 |
