siRNA Details
Gene Symbol: Srebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276707.1 | NP_001263636.1 | 2 | 288 | ||
NM_001276708.1 | NP_001263637.1 | 2 | 175 | |||
XM_008767879.1 | XP_008766101.1 | 2 | 428 | |||
GenBank mRNA | AF286469.2 | - | 2 | 142 | ||
AF286470.2 | - | 2 | 142 |
