siRNA Details
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169137.2 | NP_001162608.1 | 16 | 3189 | ||
NM_024367.1 | NP_077343.1 | 15 | 2853 | |||
XM_006250818.2 | XP_006250880.1 | 15 | 3635 | |||
XM_006250819.2 | XP_006250881.1 | 15 | 3632 | |||
XM_008770059.1 | XP_008768281.1 | 16 | 3701 | |||
GenBank mRNA | X78689.1 | - | 15 | 2920 |
