siRNA Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024394.2 | NP_077370.2 | 9 | 1317 | ||
XM_006243021.2 | XP_006243083.1 | 9 | 1346 | |||
GenBank mRNA | D49395.1 | - | 9 | 1208 | ||
DQ226540.1 | - | 2 | 268 |
