siRNA Details
Gene Symbol: Mvd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031062.2 | NP_112324.1 | 7 | 821 | ||
GenBank mRNA | BC081784.1 | - | 7 | 806 | 7135885 | |
U53706.1 | - | 7 | 822 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mvd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031062.2 | NP_112324.1 | 7 | 821 | ||
GenBank mRNA | BC081784.1 | - | 7 | 806 | 7135885 | |
U53706.1 | - | 7 | 822 |