siRNA Details
Gene Symbol: Rock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031098.1 | NP_112360.1 | 8 | 957 | ||
XM_006254400.2 | XP_006254462.1 | 8 | 1709 | |||
XM_006254401.2 | XP_006254463.1 | 8 | 1709 | |||
XM_008771957.1 | XP_008770179.1 | 8 | 1709 | |||
GenBank mRNA | FQ225745.1 | - | 9 | 1433 | ||
FQ226663.1 | - | 6 | 663 | |||
U61266.1 | - | 8 | 957 |
