siRNA Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031151.2 | NP_112413.2 | 5 | 642 | ||
GenBank mRNA | BC063165.1 | - | 5 | 642 | 6918633 | |
DQ402951.1 | - | 4 | 476 | |||
FQ213494.1 | - | 5 | 595 | |||
FQ213817.1 | - | 5 | 557 | |||
FQ215385.1 | - | 5 | 602 | |||
FQ215647.1 | - | 5 | 603 | |||
FQ216782.1 | - | 5 | 633 | |||
FQ217232.1 | - | 5 | 594 | |||
FQ224847.1 | - | 5 | 539 | |||
X04240.1 | - | 5 | 583 |
