siRNA Details
Gene Symbol: Rab11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031152.2 | NP_112414.1 | 3 | 322 | ||
GenBank mRNA | BC085727.1 | - | 3 | 322 | 7189729 | |
FQ214239.1 | - | 3 | 386 | |||
FQ220492.1 | - | 3 | 394 | |||
FQ234527.1 | - | 3 | 324 | |||
M75153.1 | - | 3 | 257 |
