siRNA Details
Gene Symbol: Sort1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031767.1 | NP_113955.1 | 13 | 1574 | ||
XM_006233194.2 | XP_006233256.1 | 13 | 2335 | |||
XM_006233195.2 | XP_006233257.1 | 13 | 2333 | |||
XM_006233196.2 | XP_006233258.1 | 13 | 2329 | |||
GenBank mRNA | AF019109.1 | - | 13 | 1287 | ||
AF023621.1 | - | 11 | 1188 |
