siRNA Details
Gene Symbol: Sort1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031767.1 | NP_113955.1 | 2, 3 | 392 | ||
XM_006233194.2 | XP_006233256.1 | 2, 3 | 1153 | |||
XM_006233195.2 | XP_006233257.1 | 2, 3 | 1154 | |||
XM_006233196.2 | XP_006233258.1 | 2, 3 | 1150 | |||
GenBank mRNA | AF019109.1 | - | 2, 3 | 105 |
