siRNA Details
Gene Symbol: Rest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031788.1 | NP_113976.1 | 3 | 2681 | ||
XM_006250847.2 | XP_006250909.1 | 2760 | ||||
XM_006250848.1 | XP_006250910.1 | 4 | 2765 | |||
XM_006250849.2 | XP_006250911.1 | 4 | 3023 | |||
GenBank mRNA | AF037199.1 | - | 3 | 2681 | ||
DQ288845.1 | - | 1 | 450 |
