siRNA Details
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031832.1 | NP_114020.1 | 2, 3 | 53 | ||
GenBank mRNA | BC089054.1 | - | 2, 3 | 41 | 7312891 | |
FQ210101.1 | - | 2, 3 | 63 | |||
FQ214799.1 | - | 2, 3 | 335 | |||
FQ218337.1 | - | 2, 3 | 53 | |||
FQ219929.1 | - | 2, 3 | 53 | |||
FQ219987.1 | - | 2, 3 | 53 | |||
FQ220157.1 | - | 2, 3 | 53 | |||
FQ220174.1 | - | 2, 3 | 52 | |||
FQ220215.1 | - | 2, 3 | 55 | |||
FQ220224.1 | - | 2, 3 | 48 | |||
FQ220313.1 | - | 2, 3 | 53 | |||
FQ220348.1 | - | 2, 3 | 53 | |||
FQ220361.1 | - | 2, 3 | 212 | |||
FQ220439.1 | - | 2, 3 | 52 | |||
FQ220450.1 | - | 2, 3 | 53 | |||
FQ220487.1 | - | 3, 4 | 253 | |||
FQ220505.1 | - | 2, 3 | 53 | |||
FQ220541.1 | - | 2, 3 | 53 | |||
FQ220563.1 | - | 2, 3 | 53 | |||
FQ220590.1 | - | 2, 3 | 55 | |||
FQ220652.1 | - | 2, 3 | 53 | |||
FQ220701.1 | - | 2, 3 | 52 | |||
FQ220837.1 | - | 2, 3 | 52 | |||
FQ220917.1 | - | 2, 3 | 53 | |||
FQ220938.1 | - | 2, 3 | 53 | |||
FQ220946.1 | - | 2, 3 | 53 | |||
FQ220952.1 | - | 2, 3 | 53 | |||
FQ221377.1 | - | 2, 3 | 53 | |||
FQ223467.1 | - | 2, 3 | 55 | |||
FQ223525.1 | - | 2, 3 | 53 | |||
FQ223531.1 | - | 2, 3 | 53 | |||
FQ229211.1 | - | 2, 3 | 57 | |||
FQ229249.1 | - | 2, 3 | 53 | |||
FQ229259.1 | - | 2, 3 | 53 | |||
FQ229266.1 | - | 2, 3 | 53 | |||
FQ229277.1 | - | 2, 3 | 50 | |||
FQ229326.1 | - | 2, 3 | 53 | |||
FQ229348.1 | - | 2, 3 | 53 | |||
FQ229362.1 | - | 2, 3 | 53 | |||
FQ229490.1 | - | 2, 3 | 53 | |||
FQ229494.1 | - | 2, 3 | 53 | |||
FQ229511.1 | - | 2, 3 | 53 | |||
FQ229560.1 | - | 2, 3 | 55 | |||
FQ229562.1 | - | 2, 3 | 55 | |||
FQ229564.1 | - | 2, 3 | 55 | |||
FQ229577.1 | - | 2, 3 | 53 | |||
FQ229661.1 | - | 2, 3 | 53 | |||
FQ229821.1 | - | 2, 3 | 53 | |||
FQ229967.1 | - | 2, 3 | 55 | |||
FQ229990.1 | - | 2, 3 | 55 | |||
FQ229991.1 | - | 2, 3 | 55 | |||
FQ229995.1 | - | 2, 3 | 53 | |||
FQ230027.1 | - | 2, 3 | 53 | |||
FQ230050.1 | - | 2, 3 | 56 | |||
FQ230072.1 | - | 2, 3 | 53 | |||
FQ230121.1 | - | 2, 3 | 60 | |||
FQ230149.1 | - | 2, 3 | 55 | |||
J02962.1 | - | 2, 3 | 53 | |||
KF639951.1 | - | 1, 2 | 13 | |||
KF639953.1 | - | 1, 2 | 13 | |||
KF639954.1 | - | 1, 2 | 13 | |||
KF639957.1 | - | 1, 2 | 13 | |||
KF639958.1 | - | 1, 2 | 13 |
