siRNA Details
Gene Symbol: Dgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053437.1 | NP_445889.1 | 2, 3 | 357 | ||
GenBank mRNA | AB062759.1 | - | 2, 3 | 335 | ||
AB062760.1 | - | 2, 3 | 335 | |||
AB062761.1 | - | 2, 3 | 335 | |||
AB062762.1 | - | 2, 3 | 335 | |||
AB062763.1 | - | 2, 3 | 333 | |||
AF296131.1 | - | 2, 3 | 357 | |||
BC081742.1 | - | 2, 3 | 329 |
