siRNA Details
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053445.2 | NP_445897.2 | 5 | 721 | ||
XM_006231075.2 | XP_006231137.1 | 5 | 868 | |||
GenBank mRNA | AB052085.1 | - | 5 | 721 | ||
AF320509.1 | - | 5 | 750 | |||
FQ218734.1 | - | 4 | 654 | |||
HQ909026.1 | - | 5 | 645 |
