siRNA Details
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053460.1 | NP_445912.1 | 2, 3 | 221 | ||
GenBank mRNA | AF327562.1 | - | 2, 3 | 221 | ||
AF373289.1 | - | 2, 3 | 143 | |||
AF542094.1 | - | 2, 3 | 191 | |||
AJ240083.2 | - | 2, 3 | 263 | |||
BC072689.1 | - | 3, 4 | 224 | 7105747 |
