siRNA Details
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053463.1 | NP_445915.1 | 10 | 1034 | ||
XM_006229163.1 | XP_006229225.1 | 11 | 1154 | |||
XM_006229164.2 | XP_006229226.1 | 11 | 1098 | |||
XM_006229165.1 | XP_006229227.1 | 11 | 1151 | |||
GenBank mRNA | AF336828.1 | - | 10 | 1034 | ||
BC100643.1 | - | 11 | 1084 | 7930904 | ||
Z36277.1 | - | 11 | 1096 |
