siRNA Details
Gene Symbol: Ppp1r9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053474.1 | NP_445926.1 | 7 | 2413 | ||
XM_008768126.1 | XP_008766348.1 | 6 | 1921 | |||
GenBank mRNA | AF016252.1 | - | 7 | 2413 | ||
BC160878.1 | - | 7 | 2095 | 7376917 |
