siRNA Details
Gene Symbol: Ppp1r9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053474.1 | NP_445926.1 | 2, 3 | 1891 | ||
XM_008768126.1 | XP_008766348.1 | 1, 2 | 1399 | |||
GenBank mRNA | AF016252.1 | - | 2, 3 | 1891 | ||
BC160878.1 | - | 2, 3 | 1573 | 7376917 |
