siRNA Details
Gene Symbol: Pola2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053480.2 | NP_445932.2 | 12, 13 | 1300 | ||
GenBank mRNA | AJ011606.1 | - | 9, 10 | 854 | ||
AJ245648.1 | - | 12, 13 | 1169 | |||
BC097300.1 | - | 12, 13 | 1300 | 7457570 |
