siRNA Details
Gene Symbol: Pik3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243642.2 | XP_006243704.1 | 11 | 1692 | ||
XM_008766567.1 | XP_008764789.1 | 6 | 1079 | |||
GenBank mRNA | AJ012482.1 | - | 11 | 1675 |
