siRNA Details
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053501.2 | NP_445953.1 | 2 | 208 | ||
XM_006257532.1 | XP_006257594.1 | 2 | 254 | |||
GenBank mRNA | AB028879.1 | - | 1 | 14 | ||
AF300424.1 | - | 2 | 209 | |||
AJ300165.1 | - | 1 | 14 |
