siRNA Details
Gene Symbol: Tekt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053508.2 | NP_445960.1 | 7 | 1108 | ||
GenBank mRNA | AJ306427.1 | - | 6 | 1037 | ||
BC078773.1 | - | 7 | 1108 | 7133616 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tekt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053508.2 | NP_445960.1 | 7 | 1108 | ||
GenBank mRNA | AJ306427.1 | - | 6 | 1037 | ||
BC078773.1 | - | 7 | 1108 | 7133616 |