siRNA Details
Gene Symbol: Tdrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138871.1 | NP_620226.1 | 16, 17 | 3250 | ||
XM_006238107.2 | XP_006238169.1 | 16, 17 | 3081 | |||
XM_006238108.2 | XP_006238170.1 | 15, 16 | 3014 | |||
XM_006238111.2 | XP_006238173.1 | 15, 16 | 3251 | |||
XM_008763704.1 | XP_008761926.1 | 15, 16 | 3408 | |||
GenBank mRNA | AB030644.1 | - | 16, 17 | 3250 | ||
BC090066.1 | - | 15, 16 | 2969 | 7122915 |
