siRNA Details
Gene Symbol: Tdrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138871.1 | NP_620226.1 | 18 | 3485 | ||
XM_006238107.2 | XP_006238169.1 | 18 | 3316 | |||
XM_006238108.2 | XP_006238170.1 | 17 | 3249 | |||
XM_006238111.2 | XP_006238173.1 | 17 | 3486 | |||
XM_008763704.1 | XP_008761926.1 | 17 | 3643 | |||
GenBank mRNA | AB030644.1 | - | 18 | 3485 | ||
BC090066.1 | - | 17 | 3204 | 7122915 |
