siRNA Details
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053527.1 | NP_445979.1 | 11 | 1608 | ||
GenBank mRNA | AF000578.2 | - | 11 | 1608 | ||
BC087007.1 | - | 12 | 1655 | |||
BC161839.1 | - | 11 | 1572 | 9101355 |
