siRNA Details
Gene Symbol: Enpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053535.1 | NP_445987.1 | 17 | 1628 | ||
XM_006227692.2 | XP_006227754.1 | 17 | 1650 | |||
GenBank mRNA | AF320054.1 | - | 17 | 1628 | ||
AF340185.1 | - | 17 | 1628 | |||
AF340186.1 | - | 17 | 1628 |
