siRNA Details
Gene Symbol: Laptm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053538.2 | NP_445990.2 | 6 | 559 | ||
GenBank mRNA | AB046592.1 | - | 6 | 571 | ||
BC060517.1 | - | 6 | 559 | 5597893 | ||
FQ225663.1 | - | 6 | 557 |
