siRNA Details
Gene Symbol: Sfrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053544.1 | NP_445996.1 | 2 | 591 | ||
GenBank mRNA | AF012891.1 | - | 2 | 591 | ||
AF140346.1 | - | 2 | 742 | |||
AF220608.1 | - | 839 | ||||
FQ229224.1 | - | 1 | 22 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
