siRNA Details
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053571.1 | NP_446023.1 | 7, 8 | 1036 | ||
XM_008769682.1 | XP_008767904.1 | 13, 14 | 4568 | |||
XM_008769683.1 | XP_008767905.1 | 11, 12 | 4276 | |||
XM_008769684.1 | XP_008767906.1 | 8, 9 | 1800 | |||
XM_008769685.1 | XP_008767907.1 | 10, 11 | 3553 | |||
XM_008769686.1 | XP_008767908.1 | 6, 7 | 837 | |||
GenBank mRNA | AB060653.1 | - | 7, 8 | 1036 |
