siRNA Details
Gene Symbol: Olfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053573.1 | NP_446025.1 | 6 | 1246 | ||
XM_006233875.2 | XP_006233937.1 | 6 | 1206 | |||
GenBank mRNA | U03416.1 | - | 6 | 1108 | ||
U03417.1 | - | 6 | 1246 |

More Information
Other Species Targeted

Gene Ontology Categories:
