siRNA Details
Gene Symbol: Cabin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053575.2 | NP_446027.2 | 25 | 4238 | ||
GenBank mRNA | AF061947.1 | - | 25 | 4244 |
Gene Symbol: LOC103689948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256337.2 | XP_006256399.1 | 1379 |
