siRNA Details
Gene Symbol: Idh3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053581.1 | NP_446033.1 | 9 | 804 | ||
XM_006235048.2 | XP_006235110.1 | 9 | 841 | |||
GenBank mRNA | AB047540.1 | - | 2 | 73 | ||
BC079113.1 | - | 9 | 804 | 7126029 | ||
FQ214820.1 | - | 9 | 802 | |||
FQ215180.1 | - | 9 | 808 | |||
FQ215184.1 | - | 9 | 796 | |||
FQ215350.1 | - | 9 | 804 | |||
FQ217667.1 | - | 3 | 162 |
