siRNA Details
Gene Symbol: Idh3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053581.1 | NP_446033.1 | 6 | 452 | ||
XM_006235048.2 | XP_006235110.1 | 6 | 489 | |||
GenBank mRNA | BC079113.1 | - | 6 | 452 | 7126029 | |
FQ214820.1 | - | 6 | 450 | |||
FQ215180.1 | - | 6 | 456 | |||
FQ215184.1 | - | 6 | 444 | |||
FQ215350.1 | - | 6 | 452 |
