siRNA Details
Gene Symbol: Clec2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130402.2 | NP_569086.1 | 3, 4 | 475 | ||
XM_008763309.1 | XP_008761531.1 | 4, 5 | 972 | |||
GenBank mRNA | AF321552.1 | - | 5, 6 | 661 | ||
EF100688.1 | - | 3, 4 | 475 |
