siRNA Details
Gene Symbol: Clec4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053753.1 | NP_446205.1 | 4 | 1074 | ||
GenBank mRNA | BC098661.1 | - | 4 | 1105 | 7384948 | |
J03734.1 | - | 4 | 1065 |

More Information
Other Species Targeted

Gene Ontology Categories:
