siRNA Details
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053760.2 | NP_446212.2 | 10 | 1630 | ||
XM_006249376.2 | XP_006249438.1 | 10 | 1686 | |||
XM_008769209.1 | XP_008767431.1 | 10 | 1733 | |||
GenBank mRNA | BC088344.1 | - | 1 | 349 |
