siRNA Details
Gene Symbol: Wnt7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100473.1 | NP_001093943.1 | 4 | 1070 | ||
GenBank mRNA | AF481945.1 | - | 1 | 205 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wnt7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100473.1 | NP_001093943.1 | 4 | 1070 | ||
GenBank mRNA | AF481945.1 | - | 1 | 205 |