siRNA Details
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105745.1 | NP_001099215.1 | 2, 3 | 182 | ||
GenBank mRNA | L23862.1 | - | 2, 3 | 152 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011139, AK153697, Z18537, L14677.
Gene Ontology Categories:
