siRNA Details
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053883.2 | NP_446335.1 | 1 | 741 | ||
GenBank mRNA | AY724528.1 | - | 1 | 742 | ||
BC087003.1 | - | 1 | 741 | 7124105 | ||
FQ217285.1 | - | 1 | 754 | |||
U42627.1 | - | 1 | 668 | |||
X94185.1 | - | 1 | 534 |
