siRNA Details
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057142.1 | NP_476483.1 | 4 | 463 | ||
GenBank mRNA | EU348640.1 | - | 3 | 386 | ||
EU348641.1 | - | 3 | 387 | |||
EU348642.1 | - | 4 | 463 | |||
EU348643.1 | - | 4 | 463 | |||
EU348644.1 | - | 4 | 463 | |||
U66707.1 | - | 4 | 463 |
