siRNA Details
Gene Symbol: Rab3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080580.2 | NP_542147.1 | 2 | 286 | ||
XM_006242593.1 | XP_006242655.1 | 2 | 282 | |||
XM_006242594.2 | XP_006242656.1 | 2 | 402 | |||
GenBank mRNA | BC081741.1 | - | 2 | 286 | 7110109 | |
M83681.1 | - | 1 | 114 | |||
S68808.1 | - | 1 | 132 | |||
U90206.1 | - | 1 | 177 |
