siRNA Details
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133394.1 | NP_596885.1 | 3 | 453 | ||
GenBank mRNA | AY040615.1 | - | 3 | 453 | ||
AY886525.1 | - | 3 | 448 | |||
BC061769.1 | - | 2 | 229 | 5621307 |
