siRNA Details
Gene Symbol: Cx3cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133534.1 | NP_598218.1 | 2 | 487 | ||
XM_008766668.1 | XP_008764890.1 | 2 | 802 | |||
GenBank mRNA | U04808.1 | - | 2 | 487 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
